Learn More
Biomatik Corporation ARL6IP5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507827
Encompass_Preferred
The ARL6IP5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human PRA1 family protein 3 protein (136-188AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: PRA1 family protein 3, ADP-ribosylation factor-like protein 6-interacting protein 5, ARL-6-interacting protein 5, Aip-5, Cytoskeleton-related vitamin A-responsive protein, Dermal papilla-derived protein 11, GTRAP3-18, Glutamate transporter EAAC1-interacting protein, JM5, Prenylated Rab acceptor protein 2, Protein JWa, Putative MAPK-activating protein PM27, ARL6IP5, DERP11, JWA, PRA2, PRAF3, HSPC127. UniProt: O75915. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7827 Supplier Biomatik Corporation Supplier No. CAC08243
The ARL6IP5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human PRA1 family protein 3 protein (136-188AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: PRA1 family protein 3, ADP-ribosylation factor-like protein 6-interacting protein 5, ARL-6-interacting protein 5, Aip-5, Cytoskeleton-related vitamin A-responsive protein, Dermal papilla-derived protein 11, GTRAP3-18, Glutamate transporter EAAC1-interacting protein, JM5, Prenylated Rab acceptor protein 2, Protein JWa, Putative MAPK-activating protein PM27, ARL6IP5, DERP11, JWA, PRA2, PRAF3, HSPC127. UniProt: O75915. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.