Learn More
Biomatik Corporation ARSB anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08244
Encompass_Preferred
The ARSB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Arylsulfatase B protein (37-533AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: N-acetylgalactosamine-4-sulfatase ARSB. UniProt: P15848. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7828