Learn More
Biomatik Corporation SRSF9 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08245
Encompass_Preferred
Srsf9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SRSF9. The Srsf9 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Arginine-Rich Splicing Factor 9 Protein (1-221Aa). Applications: ELISA, WB. Target Synonyms: SRSF9. Accession Number: Q13242. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7829