Learn More
Biomatik Corporation ARSG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08246
Encompass_Preferred
The ARSG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Arylsulfatase G protein (17-525AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: ASG, UNQ839/PRO1777, KIAA1001, ARSG. UniProt: Q96EG1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7830