Learn More
Biomatik Corporation ASCC1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08247
Encompass_Preferred
The ASCC1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Activating signal cointegrator 1 complex subunit 1 protein (1-315AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: Activating signal cointegrator 1 complex subunit 1, ASCC1, CGI-18, ASC-1 complex subunit p50, Trip4 complex subunit p50. UniProt: Q8N9N2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7831