Learn More
Biomatik Corporation CSTB anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08250
Encompass_Preferred
The CSTB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cystatin-B protein (1-98AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: CST6, STFB, CSTB, CPI-B, Liver thiol proteinase inhibitor Stefin-B. UniProt: P04080. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7834