Learn More
Biomatik Corporation ATP12A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08252
Encompass_Preferred
The ATP12A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Potassium-transporting ATPase alpha chain 2 protein (171-306AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Potassium-transporting ATPase alpha chain 2, Non-gastric H (+)/K (+) ATPase subunit alpha, Proton pump, ATP12A, ATP1AL1. UniProt: P54707. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7836