Learn More
Biomatik Corporation ATP6V1G2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08253
Encompass_Preferred
The ATP6V1G2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human V-type proton ATPase subunit G 2 protein (1-118AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: V-type proton ATPase subunit G 2, V-ATPase subunit G 2, V-ATPase 13 kDa subunit 2, Vacuolar proton pump subunit G 2, ATP6V1G2, ATP6G, ATP6G2, NG38. UniProt: O95670. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7837