Learn More
Biomatik Corporation ATXN3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507839
Encompass_Preferred
The ATXN3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Ataxin-3 protein (1-288AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Ataxin-3, Machado-Joseph disease protein 1, Spinocerebellar ataxia type 3 protein, ATXN3, ATX3, MJD, MJD1, SCA3. UniProt: P54252. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7839 Supplier Biomatik Corporation Supplier No. CAC08255
The ATXN3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Ataxin-3 protein (1-288AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Ataxin-3, Machado-Joseph disease protein 1, Spinocerebellar ataxia type 3 protein, ATXN3, ATX3, MJD, MJD1, SCA3. UniProt: P54252. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.