Learn More
Biomatik Corporation BCAT1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08257
Encompass_Preferred
The BCAT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Branched-chain-amino-acid aminotransferase, cytosolic protein (3-275AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Branched-chain-amino-acid aminotransferase, cytosolic, BCAT (c), Protein ECA39, BCAT1, BCT1, ECA39. UniProt: P54687. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7841