Learn More
Biomatik Corporation BMPR1A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08260
Encompass_Preferred
The BMPR1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Bone morphogenetic protein receptor type-1A protein (177-532AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Bone morphogenetic protein receptor type-1A, BMP type-1A receptor, BMPR-1A, Activin receptor-like kinase 3, ALK-3, Serine/threonine-protein kinase receptor R5, SKR5, BMPR1A, ACVRLK3, ALK3. UniProt: P36894. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7844