Learn More
Biomatik Corporation LGB anti-Bovine, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08262
Encompass_Preferred
Lgb Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to LGB. The Lgb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Bovine Milk Beta-Lactoglobulin (Native Protein). Applications: ELISA, WB. Target Synonyms: LGB. Accession Number: P02754. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7846