Learn More
Biomatik Corporation a1-acidglycoprotein anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08266
Encompass_Preferred
The a1-acidglycoprotein Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Human 1-acidglycoprotein (Native Protein). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: 1-AGP. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7850