Learn More
Biomatik Corporation HEATR9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08267
Encompass_Preferred
The HEATR9 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein HEATR9 protein (1-219AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Protein HEATR9, HEATR9, C17orf66, HEAT repeat-containing protein 9. UniProt: A2RTY3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7851