Learn More
Biomatik Corporation EVA1C anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08272
Encompass_Preferred
The EVA1C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein eva-1 homolog C protein (49-322AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Protein eva-1 homolog C, EVA1C, C21orf63, C21orf64, FAM176C, PRED34, UNQ2504/PRO5993, Protein FAM176C, SUE21. UniProt: P58658. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7856