Learn More
Biomatik Corporation C5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08273
Encompass_Preferred
The C5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement C5 protein (678-751AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Complement C5, C5, CPAMD4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, Complement C5 beta chain, Complement C5 alpha chain, C5a anaphylatoxin, Complement C5 alpha' chain. UniProt: P01031. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7857