Learn More
Biomatik Corporation COA1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08274
Encompass_Preferred
The COA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cytochrome c oxidase assembly factor 1 homolog protein (38-146AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cytochrome c oxidase assembly factor 1 homolog, Mitochondrial translation regulation assembly intermediate of cytochrome c oxidase protein of 15 kDa, COA1, C7orf44, MITRAC15. UniProt: Q9GZY4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7858