Learn More
Biomatik Corporation CAP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08280
Encompass_Preferred
The CAP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Adenylyl cyclase-associated protein 2 protein (221-477AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Adenylyl cyclase-associated protein 2, CAP 2, CAP2. UniProt: P40123. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7863