Learn More
Biomatik Corporation CAPN2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08282
Encompass_Preferred
The CAPN2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Calpain-2 catalytic subunit protein (20-700AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Calpain M-type, Calpain large polypeptide L2, Calpain-2 large subunit, Millimolar-calpain, CAPN2, CANPL2. UniProt: P17655. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7865