Learn More
Biomatik Corporation CARS anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507866
Encompass_Preferred
The CARS Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cysteine--tRNA ligase, cytoplasmic protein (201-500AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200. Alternative Names: Cysteine--tRNA ligase, cytoplasmic, Cysteinyl-tRNA synthetase, CysRS, CARS. UniProt: P49589. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7866 Supplier Biomatik Corporation Supplier No. CAC08283
The CARS Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cysteine--tRNA ligase, cytoplasmic protein (201-500AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:2000-1:5000, IHC:1:20-1:200. Alternative Names: Cysteine--tRNA ligase, cytoplasmic, Cysteinyl-tRNA synthetase, CysRS, CARS. UniProt: P49589. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.