Learn More
Biomatik Corporation HSBP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08284
Encompass_Preferred
The HSBP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Heat shock factor-binding protein 1 protein (1-75AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Nasopharyngeal carcinoma-associated antigen 13, HSBP1, HSF1BP. UniProt: O75506. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7867