Learn More
Biomatik Corporation CD7 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08296
Encompass_Preferred
The CD7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human T-cell antigen CD7 protein (26-180AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: GP40, T-cell leukemia antigen, T-cell surface antigen Leu-9, TP41, CD7. UniProt: P09564. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7879