Learn More
Biomatik Corporation Cd74 anti-Rat, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08297
Encompass_Preferred
The Cd74 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Mouse. Immunogen: Recombinant rat H-2 class II histocompatibility antigen gamma chain protein (57-280AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Ia antigen-associated invariant chain, Ii, MHC class II-associated invariant chain, CD74, Cd74. UniProt: P10247. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7880