Learn More
Biomatik Corporation BLOC1S1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08298
Encompass_Preferred
The BLOC1S1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Biogenesis of lysosome-related organelles complex 1 subunit 1 (1-125AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GCN5-like protein 1 Protein RT14. UniProt: P78537. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7881