Learn More
Biomatik Corporation Cd82 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08299
Encompass_Preferred
The Cd82 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse CD82 antigen protein (111-227AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: C33 antigen, IA4, Inducible membrane protein R2, Metastasis suppressor Kangai-1 homolog, CD82, Cd82, Kai1. UniProt: P40237. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7882