Learn More
Biomatik Corporation CDH11 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08301
Encompass_Preferred
The CDH11 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Cadherin-11 protein (637-796AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cadherin-11, OSF-4, Osteoblast cadherin, OB-cadherin, CDH11. UniProt: P55287. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7884