Learn More
Biomatik Corporation CDKN1A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08304
Encompass_Preferred
The CDKN1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Cyclin-dependent kinase inhibitor 1 protein (2-164AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:1000-1:2000, IF:1:200-1:500. Alternative Names: CDK-interacting protein 1, Melanoma differentiation-associated protein 6, p21, CDKN1A, CAP20, CDKN1, CIP1, MDA6, PIC1, SDI1, WAF1. UniProt: P38936. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7887