Learn More
Biomatik Corporation CDKN2B anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507888
Encompass_Preferred
The CDKN2B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cyclin-dependent kinase 4 inhibitor B protein (1-138AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cyclin-dependent kinase 4 inhibitor B, Multiple tumor suppressor 2, MTS-2, p14-INK4b, p15-INK4b, p15INK4B, CDKN2B, MTS2. UniProt: P42772. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7888 Supplier Biomatik Corporation Supplier No. CAC08305
The CDKN2B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cyclin-dependent kinase 4 inhibitor B protein (1-138AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cyclin-dependent kinase 4 inhibitor B, Multiple tumor suppressor 2, MTS-2, p14-INK4b, p15-INK4b, p15INK4B, CDKN2B, MTS2. UniProt: P42772. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.