Learn More
Biomatik Corporation CFLAR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08309
Encompass_Preferred
The CFLAR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CASP8 and FADD-like apoptosis regulator protein (1-250AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:200-1:500. Alternative Names: CASP8 and FADD-like apoptosis regulator, CFLAR, CASH, CASP8AP1, CLARP, MRIT, Caspase homolog, CASH, Caspase-eight-related protein, Casper, Caspase-like apoptosis regulatory protein, CLARP, Cellular FLICE-like inhibitory protein, c-FLIP, FADD-like antiapoptotic molecule 1, FLAME-1, Inhibitor of FLICE, I-FLICE, MACH-related inducer of toxicity, MRIT, Usurpin, CASP8 and FADD-like apoptosis regulator subunit p43, CASP8 and FADD-like apoptosis regulator subunit p12. UniProt: O15519. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7892