Learn More
Biomatik Corporation CHGA anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08310
Encompass_Preferred
The CHGA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Chromogranin-A protein (19-457AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: CgA, Pituitary secretory protein I, SP-I, CHGA. UniProt: P10645. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7893