Learn More
Biomatik Corporation CHM anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08312
Encompass_Preferred
The CHM Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Rab proteins geranylgeranyltransferase component A 1 protein (1-653AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:50-1:200. Alternative Names: Rab proteins geranylgeranyltransferase component A 1, Choroideremia protein, Rab escort protein 1, REP-1, TCD protein, CHM, REP1, TCD. UniProt: P24386. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7895