Learn More
Biomatik Corporation CLASP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08313
Encompass_Preferred
The CLASP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CLIP-associating protein 2 protein (1-431AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: CLIP-associating protein 2, CLASP2, KIAA0627, Cytoplasmic linker-associated protein 2, Protein Orbit homolog 2, hOrbit2. UniProt: O75122. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7896