Learn More
Biomatik Corporation CLEC18A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08317
Encompass_Preferred
The CLEC18A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human C-type lectin domain family 18 member A protein (27-446AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Mannose receptor-like protein 2 CLEC18A MRLP2. UniProt: A5D8T8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7900