Learn More
Biomatik Corporation CLEC3A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08318
Encompass_Preferred
The CLEC3A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human C-type lectin domain family 3 member A protein (23-197AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: C-type lectin domain family 3 member A, C-type lectin superfamily member 1, Cartilage-derived C-type lectin, CLEC3A, CLECSF1, UNQ700/PRO1345. UniProt: O75596. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7901