Learn More
Biomatik Corporation CLIC2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08320
Encompass_Preferred
The CLIC2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Chloride intracellular channel protein 2 protein (53-247AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Chloride intracellular channel protein 2, XAP121, CLIC2. UniProt: O15247. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7903