Learn More
Biomatik Corporation CLTCL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08321
Encompass_Preferred
The CLTCL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Clathrin heavy chain 2 protein (1423-1566AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Clathrin heavy chain on chromosome 22, CLH-22, CLTCL1, CLH22, CLTCL, CLTD. UniProt: P53675. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7904