Learn More
Biomatik Corporation RPL14 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08322
Encompass_Preferred
Rpl14 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPL14. The Rpl14 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 60S Ribosomal Protein L14 Protein (2-214Aa). Applications: ELISA, IHC. Target Synonyms: RPL14. Accession Number: P50914. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7905