Learn More
Biomatik Corporation CNOT1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507906
Encompass_Preferred
The CNOT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CCR4-NOT transcription complex subunit 1 protein (604-867AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: CCR4-NOT transcription complex subunit 1, CCR4-associated factor 1, Negative regulator of transcription subunit 1 homolog, NOT1H, hNOT1, CNOT1, CDC39, KIAA1007, NOT1, AD-005. UniProt: A5YKK6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7906 Supplier Biomatik Corporation Supplier No. CAC08323
The CNOT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CCR4-NOT transcription complex subunit 1 protein (604-867AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: CCR4-NOT transcription complex subunit 1, CCR4-associated factor 1, Negative regulator of transcription subunit 1 homolog, NOT1H, hNOT1, CNOT1, CDC39, KIAA1007, NOT1, AD-005. UniProt: A5YKK6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.