Learn More
Biomatik Corporation Col1a1 anti-Rat, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08324
Encompass_Preferred
The Col1a1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Human. Immunogen: Recombinant rat Collagen alpha-1 (I) chain protein (955-1207AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Alpha-1 type I collagen Col1a1. UniProt: P02454. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7907