Learn More
Biomatik Corporation COL8A1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08325
Encompass_Preferred
The COL8A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Collagen alpha-1 (VIII) chain protein (572-744AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:2000-1:10000, IF:1:50-1:500. Alternative Names: Collagen alpha-1 (VIII) chain, Endothelial collagen, COL8A1, C3orf7. UniProt: P27658. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7908