Learn More
Biomatik Corporation EMC8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08327
Encompass_Preferred
The EMC8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ER membrane protein complex subunit 8 protein (1-210AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: ER membrane protein complex subunit 8, EMC8, C16orf2, C16orf4, COX4AL, COX4NB, FAM158B, NOC4, Neighbor of COX4, Protein FAM158B. UniProt: O43402. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7910
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.