Learn More
Biomatik Corporation CPA2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08329
Encompass_Preferred
The CPA2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Carboxypeptidase A2 protein (115-419AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Carboxypeptidase A2 (EC:3.4.17.15), CPA2. UniProt: P48052. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7912