Learn More
Biomatik Corporation CPLX1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08330
Encompass_Preferred
The CPLX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complexin-1 protein (1-134AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Complexin-1, Complexin I, CPX I, Synaphin-2, CPLX1. UniProt: O14810. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7913