Learn More
Biomatik Corporation CRABP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08331
Encompass_Preferred
The CRABP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cellular retinoic acid-binding protein 1 protein (2-137AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Cellular retinoic acid-binding protein I, CRABP-I, CRABP1, RBP5. UniProt: P29762. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7914