Learn More
Biomatik Corporation CRADD anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08332
Encompass_Preferred
The CRADD Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Death domain-containing protein CRADD protein (1-199AA). Applications: ELISA, IF. Recommended dilution: IF:1:200-600. Alternative Names: Caspase and RIP adapter with death domain, RIP-associated protein with a death domain, CRADD, RAIDD. UniProt: P78560. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7915