Learn More
Biomatik Corporation RPL8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08333
Encompass_Preferred
Rpl8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPL8. The Rpl8 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 60S Ribosomal Protein L8 Protein (3-257Aa). Applications: ELISA, WB. Target Synonyms: RPL8. Accession Number: P62917. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7916