Learn More
Biomatik Corporation CRNN anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507918
Encompass_Preferred
The CRNN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Cornulin protein (139-389AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Cornulin, CRNN, C1orf10, DRC1, PDRC1, SEP53, 53 kDa putative calcium-binding protein, 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, Squamous epithelial heat shock protein 53, Tumor-related protein. UniProt: Q9UBG3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7918 Supplier Biomatik Corporation Supplier No. CAC08335
The CRNN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Cornulin protein (139-389AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Cornulin, CRNN, C1orf10, DRC1, PDRC1, SEP53, 53 kDa putative calcium-binding protein, 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, Squamous epithelial heat shock protein 53, Tumor-related protein. UniProt: Q9UBG3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.