Learn More
Biomatik Corporation CRYBA1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08336
Encompass_Preferred
The CRYBA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Beta-crystallin A3 protein (1-215AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Beta-crystallin A3Cleaved into the following 3 chains:Beta-crystallin A3, isoform A1, Delta4 formBeta-crystallin A3, isoform A1, Delta7 formBeta-crystallin A3, isoform A1, Delta8 form, CRYBA1, CRYB1. UniProt: P05813. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7919