Learn More
Biomatik Corporation CRYZL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507920
Encompass_Preferred
The CRYZL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Quinone oxidoreductase-like protein 1 protein (1-87AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Quinone oxidoreductase-like protein 1 (EC:1.-.-.-), CRYZL1, 4P11, Protein 4P11, Quinone oxidoreductase homolog 1, QOH-1, Zeta-crystallin homolog. UniProt: O95825. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7920 Supplier Biomatik Corporation Supplier No. CAC08337
The CRYZL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Quinone oxidoreductase-like protein 1 protein (1-87AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Quinone oxidoreductase-like protein 1 (EC:1.-.-.-), CRYZL1, 4P11, Protein 4P11, Quinone oxidoreductase homolog 1, QOH-1, Zeta-crystallin homolog. UniProt: O95825. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.