Learn More
Biomatik Corporation CSGALNACT2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507921
Encompass_Preferred
The CSGALNACT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Chondroitin sulfate N-acetylgalactosaminyltransferase 2 protein (33-270AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Chondroitin sulfate N-acetylgalactosaminyltransferase 2 (EC:2.4.1.174), CSGALNACT2, CHGN2, GALNACT2, PRO0082, Chondroitin beta-1, 4-N-acetylgalactosaminyltransferase 2, Beta4GalNAcT-2, GalNAcT-2. UniProt: Q8N6G5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7921 Supplier Biomatik Corporation Supplier No. CAC08338
The CSGALNACT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Chondroitin sulfate N-acetylgalactosaminyltransferase 2 protein (33-270AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Chondroitin sulfate N-acetylgalactosaminyltransferase 2 (EC:2.4.1.174), CSGALNACT2, CHGN2, GALNACT2, PRO0082, Chondroitin beta-1, 4-N-acetylgalactosaminyltransferase 2, Beta4GalNAcT-2, GalNAcT-2. UniProt: Q8N6G5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.